missing translation for 'onlineSavingsMsg'
Learn More

ibidi μ-Slide 2 Well ibiTreat

Product Code. 30130521
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
90/Cs.
Unit Size:
Pack of 120
Pack of 90
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case Unit Size
30130521 90/Cs. Pack of 90
30130545 120/Cs. Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30130521 Supplier ibidi Supplier No. 8028690
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell culture slide with 2 wells for immuno-fluorescence and high-end microscopy.

  • All-in-one 2 well chamber for IF stainings and brilliant imaging in the same microscopy chamber
  • Bottom material: No. 1.5 Polymer Coverslip
  • Tissue culture treated
  • Sterilized

Specifications

Type Chamber Slide
Wells 2
Surface Treatment ibiTreat
Material Polymer
For Use With (Application) Cell Culture
No. per Case 90/Cs.
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.