Learn More
ibidi μ-Slide 2 Well Untreated
Product Code. 31941029
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
90/Cs.
No. per Pack:
1 Ea.
8/Tray, 15 Trays
Unit Size:
Pack of 120
Pack of 90
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case No. per Pack Unit Size
31941029 90/Cs. 1 Ea. Pack of 90
31941055 120/Cs. 8/Tray, 15 Trays Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31941029 Supplier ibidi Supplier No. 8028190
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cell culture slide with 2 wells for immuno-fluorescence and high-end microscopy.
  • All-in-one 2 well chamber for IF stainings and brilliant imaging in the same microscopy chamber
  • No.1.5 polymer coverslip
  • Hydrophobic
  • Sterilized
Specifications
Type Chamber Slide
Wells 2
Surface Treatment Untreated
Material Polymer
No. per Case 90/Cs.
No. per Pack 1 Ea.
Sterility Sterile