Learn More
Ibidi μ-Slide 8 Well High Collagen I
Product Code. 17454182
Encompass_Preferred
Change view
Click to view available options
Quantity:
15
Unit Size:
Pack of 15
Product Code. Quantity Unit Size
17454182 15 Pack of 15
1 options
Product Code. 17454182 Supplier Ibidi Supplier No. 80809
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide 8 Well High Collagen I is a chambered coverslip with 8 individual wells and high walls.
  • A chambered coverslip with 8 individual wells and high walls for cell culture, immunofluorescence, and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • Collagen I coated
  • sterilized
Specifications
Type Microscope Slide
Well Count 8
Quantity 15