Learn More
Ibidi μ-Slide 8 Well High Uncoated
Click to view available options
:
μ-Slide
Description
- A chambered coverslip with 8 individual wells and high walls for cell culture, immunofluorescence, and high-end microscopy
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Specifications
Specifications
| Type | Chambered Coverglass |
| Wells | 8 |