missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Device, 7K MWCO

Product Code. 10549473 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 Devices
50 Devices
Unit Size:
Pack of 250
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10549473 250 Devices Pack of 250
10083213 50 Devices Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10549473 Supplier Thermo Scientific™ Supplier No. 69562
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform sample buffer exchange with these 7000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

Slide-A-Lyzer MINI Dialysis Devices, 7K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules larger than 7,000 Daltons in volumes from 10 to 100 μL.

Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

Slide-A-Lyzer MINI Dialysis Devices allow easy sample addition and removal using a standard laboratory pipette and can be used for single or arrays of samples. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with common chemicals and buffers.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker.

Features of Slide-A-Lyzer MINI Dialysis Devices, 7K MWCO:
• Flexible format—enables user to dialyze a single sample (tube) or multiple samples (25 array)
• Excellent sample recovery—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
• One-step protocol—does not require laborious procedures or expensive equipment
• 100% leak-tested—innovative design does not permit 'wicking' that can occur in home-made devices
• Time saving—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

Using the 0.1 mL MINI Device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cup, Microcentrifuge
Product Line Slide-A-Lyzer
MWCO 7.0 kDa
Membrane Cellulose Membrane, Dialysis
Format MINI Dialysis Device
Description MINI Dialysis Device, 0.1 mL
Purification Target Buffer Exchange, Protein
Sample Volume (Metric) 0.1 mL
Quantity 250 Devices
Content And Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.