missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Flasks

Product Code. 13414209 Shop All Thermo Scientific Products
Change view
Click to view available options
MWCO:
10 kDa
2.0 kDa
20 kDa
3.5 kDa
Unit Size:
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO unitSize
13414209 20 kDa Pack of 4
13484199 2.0 kDa Pack of 4
13494199 3.5 kDa Pack of 4
13404209 10 kDa Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13414209 Supplier Thermo Scientific™ Supplier No. 87763

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Facilitate removal of salts and small contaminants from proteins or macromolecules in sample volumes up to 250 mL. Each Slide-A-Lyzer Dialysis Flask provides flotation disks that when attached to the neck of the flask, allows the flask to float in a vertical position for simple and safe handling.

Slide-A-Lyzer Dialysis Flasks (2K, 3.5K, 10K, or 20K MWCO) facilitate simple and effective removal of buffer salts and small contaminants in sample volumes up to 250 mL. They are easy to use: add and remove sample using a serological pipette or by directly pouring the sample through a wide screw-cap opening at the top of the flask. No knots or clips are needed to seal the units. Slide-A-Lyzer Dialysis Flasks are manufactured using clean room conditions and have low-protein binding regenerated cellulose membranes to ensure maximum sample recovery and sample purity.

The dialysis flasks are available in distinct colors, corresponding to the pore size (MWCO) of the dialysis membrane: purple (20,000 Daltons), orange (10,000 Daltons), pink (3,500 Daltons), and blue (2,000 Daltons).

Slide-A-Lyzer Dialysis Flasks are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of an inert plastic body. The membrane and flask materials are compatible with most laboratory buffers.

Features of Slide-A-Lyzer Dialysis Flasks:

  • Easy to use—simply pipette or pour sample into flask and begin dialysis
  • Fast dialysis—flat flask chamber with two membranes provides high surface area to volume ratio, enabling dialysis of a 250 mL sample in 8 hours to overnight
  • High recovery—rectangular flask design maximizes recovery of entire sample volume via opening at top of flask
  • Multiple molecular-weight cut offs—select the membrane MWCO that best suits your sample's molecular weight

Dialysis membrane specifications:

  • Composition: regenerated-cellulose
  • Thickness: 0.9–1.2 mil (22.5 to 30 μm); 2K MWCO = 2 mil (50 μm)
  • Hydration time required before use: 2 minutes
  • Glycerol content: trace (10K MWCO content is 21%)
  • Sulfur content: 0.04–0.17%
  • Heavy metals content: trace

With Slide-A-Lyzer Dialysis Flasks, typical low-molecular weight contaminant removal, buffer exchange, and desalting can be accomplished in eight hours to overnight. The rate of dialysis depends on a variety of factors, including sample volume, size and shape of the molecule being dialyzed, agitation, and temperature. Minimum dialysis time must be determined empirically for each specific system.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Shipped at ambient temperature. Upon receipt, store at room temperature.
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Flask
MWCO 20 kDa
Product Line Slide-A-Lyzer
Quantity 4 Flasks
Sample Volume (Metric) 250 mL
Product Type Dialysis Flasks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.