missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 3.5K MWCO

Product Code. 10615655 Shop All Thermo Scientific Products
Change view
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
15 mL
30 mL
70 mL
Unit Size:
Pack of 10
Pack of 6
Pack of 8
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Volume (Metric) Unit Size
10615655 0.5 mL Pack of 10
10003594 3 mL Pack of 10
10515104 15 mL Pack of 8
10451375 30 mL Pack of 6
10378974 70 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10615655 Supplier Thermo Scientific™ Supplier No. 87722

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation self-floating protein cassettes.

Slide-A-Lyzer G2 Dialysis Cassettes provide maximum convenience, flexibility, and performance for sample dialysis. These 3.5K molecular weight cut off (3.5K MWCO) cassettes use a cellulose membrane to retain proteins and other macromolecules larger than 3,500 Daltons while allowing removal of salts and small contaminants. The cassette membrane is composed of low-binding regenerated cellulose. The membranes and cassettes are manufactured using clean-room conditions to ensure they are contaminant-free.

Slide-A-Lyzer G2 Dialysis Cassettes dialyze sample volumes up to 70 mL. These 3.5K MWCO cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.

Features of Slide-A-Lyzer G2 Dialysis Cassettes (3.5K MWCO):
• Easy-loading—pipette-accessible for easy sample loading and retrieval
• Self-floating—integrated air chambers eliminate the need for float buoys
• Sturdy construction—ensures the highest possible sample integrity and protection
• Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
• Versatile—available for volumes of 0.5 to 70 mL

Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL

Order Info

Important Note: SKU #87722 will be discontinued and replaced by 66333, 66335, or 88403 by June 1, 2023. Consider switching to 66333, 66335, or 88403.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 3.5 kDa
Product Line Slide-A-Lyzer
Quantity 10 Cassettes
Sample Volume (Metric) 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.