Learn More
Ibidi μ-Slide Chemotaxis Collagen IV
Product Code. 17206173
Encompass_Preferred
Change view
Click to view available options
Quantity:
10
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
17206173 10 Pack of 10
1 options
Product Code. 17206173 Supplier Ibidi Supplier No. 80322
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide Chemotaxis Collagen IV is a ready-to-use microscopy slide for chemotaxis assays.
  • A ready-to-use microscopy slide for chemotaxis assays with fast or slow migrating cells
  • suitable for 2D or 3D cell culture
  • stable chemotactic gradients for long-term experiments
  • bottom material: #1.5 Polymer Coverslip
  • Collagen IV coated
  • sterilized
Specifications
Type Microscope Slide
Quantity 10