Learn More
Leica Microsystems Darkfield Slider
Product Code. 11954759
Click to view available options
Type:
Simple Darkfield Slider
Unit Size:
Each
Product Code. 11954759
Brand: Leica Microsystems 11501158
This item requires a price update prior to being ordered.
Request a Price for this item here.
Improve unstained sample image clarity by increasing sample to background contrast via darkfield methodology. Leica Microsystems™ Darkfield Slider permits visualization of small structures typically overlooked by brightfield observation.
  • Improves clarity.
  • Increases contrast.
  • Fits DM model microscopes with slotted Abbe condensers.
Specifications
Type Simple Darkfield Slider
For Use With (Equipment) DM Series microscopes with slotted Abbe condensers