Learn More
Ibidi μ-Slide I 0.2 Luer Glass Bottom
Product Code. 17206153
Encompass_Preferred
Change view
Click to view available options
μ-Slide
Product Code.
17206153 μ-Slide
1 options
Product Code. 17206153 Supplier Ibidi Supplier No. 80167
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A μ-Slide has one channel for applying defined shear stress.
  • A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
  • easy to connect with tubing and pump systems via female Luers
  • channel height: 200 μm
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • sterilized
Specifications
Type Microscope Slide
Quantity 15