Learn More
ibidi μ-Slide I 0.2 Luer Uncoated
Product Code. 16281011
Encompass_Preferred
Change view
Click to view available options
15 Pk.
Product Code.
16281011 15 Pk.
1 options
Product Code. 16281011 Supplier ibidi Supplier No. 80161
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A μ-Slide with one channel
  • A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
  • Easy to connect with tubing and pump systems via female Luers
  • Channel height: 200 μm, bottom material: #1.5 Polymer Coverslip
  • Hydrophobic
  • Sterilized
Specifications
Quantity 15 Pk.