Learn More
Ibidi μ-Slide I Luer Variety Pack Uncoated
Product Code. 17266213
Encompass_Preferred
Change view
Click to view available options
μ-Slide
Product Code.
17266213 μ-Slide
1 options
Product Code. 17266213 Supplier Ibidi Supplier No. 81101
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide I Luer Variety Pack Uncoated is a variety of μ-Slide I Luer channel slides.
  • Variety of μ-Slide I Luer channel slides with 4 times each channel height
  • specially suited for optimizing experimental conditions in flow applications
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Microscope Slide
Quantity 16