Learn More
Ibidi μ-Slide III 3in1 ibiTreat
Product Code. 17296163
Encompass_Preferred
Change view
Click to view available options
μ-Slide
Product Code.
17296163 μ-Slide
1 options
Product Code. 17296163 Supplier Ibidi Supplier No. 80316
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide III 3in1 ibiTreat is a μ-Slide for switchable chemotactic gradients.
  • A channel slide for switchable chemotactic gradients that merges 3 separated liquids into one channel
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Type Microscope Slide
Quantity 15