Learn More
Ibidi μ-Slide Spheroid Perfusion Bioinert
Product Code. 17226173
Encompass_Preferred
Change view
Click to view available options
μ-Slide
Product Code.
17226173 μ-Slide
1 options
Product Code. 17226173 Supplier Ibidi Supplier No. 80350
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide Spheroid Perfusion Bioinert is a perfusable channel slide with 3 x 7 wells.
  • A perfusable channel slide with 3 x 7 wells for long-term spheroid cultivation and high-end microscopy
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert)
  • sterilized
Specifications
Type Microscope Slide
Quantity 15