Learn More
ibidi μ-Slide Spheroid Perfusion ibiTreat
Product Code. 30130494
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
90/Cs.
No. per Pack:
1 Ea.
8/Tray, 15 Trays
Unit Size:
Pack of 120
Pack of 90
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case No. per Pack Unit Size
30130494 90/Cs. 1 Ea. Pack of 90
30130571 120/Cs. 8/Tray, 15 Trays Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30130494 Supplier ibidi Supplier No. 8035690
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A perfusable channel μ-Slide with 3 x 7 wells for spheroid culture and high-end imaging.
  • Easy to connect with tubing and pump systems via female Luers
  • Bottom material: No. 1.5 Polymer Coverslip
  • Tissue culture treated
  • Sterilized
  • Ideal for the long-term cultivation and perfusion of 3D spheroids or organoids on the Bioinert surface
  • Flexible preparation for perfusion: spheroids can be transferred or created directly in the wells
  • Imaging chamber with excellent optical quality for high-resolution microscopy
Specifications
Type Chamber Slide
Wells 7
Surface Treatment ibiTreat
Material Polymer
No. per Case 90/Cs.
No. per Pack 1 Ea.
Sterility Sterile