Learn More
BRAND™ Slide Staining Trough
Product Code. 10251951 Shop All Brand Products
Change view
Click to view available options
Material:
POM
Unit Size:
Pack of 5
Product Code. Material Unit Size
10251951 POM Pack of 5
1 options
Product Code. 10251951 Supplier BRAND™ Supplier No. 471500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Slide Staining Trough with a close fitting lid that helps protect against evaporation.
  • For 25 slides
  • Close fitting lid helps protect against evaporation
  • Tray has a fold down handle
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Slides
Type Staining Trough
Material POM