Learn More
Ibidi μ-Slide VI 0.4 Collagen I
Product Code. 17854834
Encompass_Preferred
Change view
Click to view available options
Quantity:
15
Unit Size:
Pack of 15
Product Code. Quantity Unit Size
17854834 15 Pack of 15
1 options
Product Code. 17854834 Supplier Ibidi Supplier No. 80609
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide VI 0.4 Collagen I is a 6 channel slide for parallel flow assays and immunofluorescence.
  • A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
  • bottom material: #1.5 Polymer Coverslip
  • Collagen I coated
  • sterilized
Specifications
Type Microscope Slide
Quantity 15