Learn More
Ibidi μ-Slide VI 0.4 Poly-L-Lysine
Product Code. 17226193
Encompass_Preferred
Change view
Click to view available options
Quantity:
15
Unit Size:
Pack of 15
Product Code. Quantity Unit Size
17226193 15 Pack of 15
1 options
Product Code. 17226193 Supplier Ibidi Supplier No. 80604
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide VI 0.4 Poly-L-Lysine is a 6 channel slide for parallel flow assays and immunofluorescence.
  • A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
  • bottom material: #1.5 Polymer Coverslip
  • Poly-L-Lysine coated
  • sterilized
Specifications
Type Microscope Slide
Quantity 15