Learn More
Ibidi μ-Slide VI - Flat ibiTreat
Product Code. 17266193
Encompass_Preferred
Change view
Click to view available options
Quantity:
15
Unit Size:
Pack of 15
Product Code. Quantity Unit Size
17266193 15 Pack of 15
1 options
Product Code. 17266193 Supplier Ibidi Supplier No. 80626
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide VI - Flat ibiTreat is a 6 channel slide for short term assays.
  • A 6 channel slide for short term assays
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Type Microscope Slide
Quantity 15