Learn More
Ibidi μ-Slide VI - Flat Uncoated
Product Code. 17256193
Encompass_Preferred
Change view
Click to view available options
μ-Slide
Product Code.
17256193 μ-Slide
1 options
Product Code. 17256193 Supplier Ibidi Supplier No. 80621
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide VI - Flat Uncoated is a 6 channel slide for short term assays.
  • A 6 channel slide for short term assays
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Microscope Slide
Quantity 15