missing translation for 'onlineSavingsMsg'
Learn More

Manutan Sliding door mesh cabinet, green, 1830mm x 1220mm x 460mm

Product Code. 12871059
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12871059 Supplier Manutan Supplier No. A343165

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A range of cabinets suitable for most work environments giving visibility of stock whilst maintaining security. Welded construction from 0. 9mm thick steel with reinforced mesh panel doors providing visibility and ventilation. Adjustable shelves, locks and keys supplied.

  • Green door
  • Three shelves
  • Dimensions (H x W x D) : 1830mm x 1220mm x 460mm
  • Capacity : 75kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.