Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for four inner-door, ultra-low temperature freezers.
Brand:  Thermo Scientific™ 1950592
Code : 51
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11860543
  • 240.00 € / Each
In Stock
to see stock.
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 4 shelf Series Freezers as well as well as previous generation Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
Specifications
Specifications
Side Access Rack (Microplate/Deepwell)
Silver
Stainless Steel
11
14 cm
30.0 cm
25.7 in.
Side Access Microplate Rack - Standard or Deepwell
Microplates
147 Plates/Rack
Thermo Scientific TDE Series and TSX Series (4 shelves)
5.5 in.
11.8 in.
65.3 cm