Learn More
Syngene™ Slimline Flat White Light Panel - Fold Down
Product Code. 13191514
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
Product Code. Quantity Unit Size
13191514 1 Each
1 options
Product Code. 13191514 Supplier Syngene™ Supplier No. WLP2014GX
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Syngene™ Slimline Flat White Light Panel - Fold Down is for use with G:BOX F3 and G:BOX Chemi XRQ systems
  •  Size: 20 x 14cm
  • Fold down white light pad with brackets
  • Includes a fold down tray, built into a standard size G:BOX darkroom cabinet.
Specifications
For Use With (Equipment) Syngene Gbox F3 and XRQ
Quantity 1