missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Push-On Lid for Urine Beaker, PE, Red, CE-IVD

Product Code. 10009710 Shop All Brand Products
Change view
Click to view available options
No. per Pack:
1000/Pk.
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
10009710 1000/Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10009710 Supplier Brand™ Supplier No. 758902
  • ONLINE EXCLUSIVE PRICE
    84.55 €
    65.90 € / Pack of 1000
    Save 18.65 €
    22% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from Polypropylene. BRAND™ Press-on Caps for Urine Beakers provide hygienic and odorless urine sample handling. To use, simply cut off integrated spout tip and insert test strip.

  • PP, snap cap PE
  • For clean and hygienic handling of urine samples without odor
  • The lid closes by simply pressing on and does not need to be opened in the laboratory
  • Simply cut the protusion in the lid and insert test strips or pour out the contents
  • CE marked according to guideline EU IVDR 2017/746
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lid
Closure Type Press-on Cap
Material Polyethylene
Closure Size 70 mm
Color Red
For Use With (Application) For Hygienic and Odorless Urine Sample Handling
No. per Pack 1000/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.