missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SLM 6031434 hydrochloride
GREENER_CHOICE

Product Code. 18734908
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18734908

Brand: Tocris Bioscience™ 6281/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective sphingosine kinase 2 (Sphk2) inhibitor

SLM 6031434 hydrochloride is a Sphk2 inhibitor (Ki = 0.4 μM for mouse Sphk2). Exhibits 40-fold selectivity for Sphk2 over Sphk1. Decreases levels of sphingosine-1-phosphate (S1P) in U937 cells in vitro and increases blood levels of S1P in wild-type mice .
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-2-[3-[4-(Octyloxy)-3-(trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl]-1-pyrrolidinecarboximidamide hydrochloride
CAS 1897379-34-8
Quantity 10 mg
Molecular Formula C22H30F3N5O2.HCl
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.