missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SLM 6031434 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SLM 6031434 hydrochloride is a Sphk2 inhibitor (Ki = 0.4 μM for mouse Sphk2). Exhibits 40-fold selectivity for Sphk2 over Sphk1. Decreases levels of sphingosine-1-phosphate (S1P) in U937 cells in vitro and increases blood levels of S1P in wild-type mice .
Specifications
Specifications
| Chemical Name or Material | (2S)-2-[3-[4-(Octyloxy)-3-(trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl]-1-pyrrolidinecarboximidamide hydrochloride |
| CAS | 1897379-34-8 |
| Quantity | 10 mg |
| Molecular Formula | C22H30F3N5O2.HCl |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction