Learn More
R&D Systems™ SM 102
Product Code. 30537733
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30537733 25 mg 25mg
1 options
Product Code. 30537733 Supplier R&D Systems™ Supplier No. 8909/25
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Tocris product, part of R&D Systems. Cationic ionizable lipid
SM 102 is an ionizable cationic lipid (pKa = 6.68) used in the formulation of lipid nanoparticles (LPNs) for the delivery of mRNA in vitro and in vivo. Administration of LNPs containing SM 102 induces hepatic luciferase expression in mice. SM 102 is commonly formulated with cholesterol (Cat. No. 7945), DSPC (Cat. No. 7943), and DMG-PEG 2000 (Cat. No. 7944).
For more information on LNPs and available components see our Lipid Nanoparticles page .
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C
Product Line Tocris
Quantity 25 mg