missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SM 324405
GREENER_CHOICE

Product Code. p-7110826
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18700949

Brand: Tocris Bioscience™ 5445/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TLR7 agonist

SM 324405 is a potent TLR7 agonist (EC50 = 50 nM). Exhibits selectivity for TLR7 over TLR8. Induces IFN-α and -γ expression in human PBMCs and mouse splenocytes, and inhibits IL-5 production in human PBMCs in vitro. Rapidly metabolized to acid metabolite (t½ = 2.6 min in human plasma).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3-[(6-amino-2-butoxy-7,8-dihydro-8-oxo-9H-purin-9-yl)methyl]benzeneacetate
CAS 677773-91-0
Quantity 10 mg
Target Toll-like Receptor Agonists
Molecular Formula C19H23N5O4
Formula Weight 385.42
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.