Learn More
Abnova™ SMAD4/CEN18q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0234.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16043385
  • 1475.00 € / 100µL
Estimated Shipment: 19-06-2024
to see stock.
Specifications
Store at 4°C in the dark.
Tube
4089
100 μL
Liquid
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe