missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Small Core Pre-printed GHS CLP Labels

Product Code. 30041449 Shop All Brady Products
Change view
Click to view available options
Display Text:
1 Red Diamond per Sticker
2 Horizontal Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamons per Sticker
2 Stacked Red Diamonds per Sticker
3 Red Diamonds per Sticker
3 Red Diamonds per Sticker in Rotated Pyramidal Shape
3 Stacked Red Diamonds in Pyramid Formation per Sticker
4 Red Diamonds in Square Formation per Sticker
Width (Metric) Tape:
55 mm
55.00 mm
75.00 mm
105.00 mm
Material:
Paper
Polypropylene
Förpackningsstorlek:
Pack of 200
Pack of 300
Pack of 400
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Display Text Width (Metric) Tape Material Förpackningsstorlek
30041449 1 Red Diamond per Sticker 55.00 mm Polypropylene Pack of 400
30041431 3 Red Diamonds per Sticker 75.00 mm Polypropylene Pack of 300
30041410 2 Stacked Red Diamonds per Sticker 105.00 mm Paper Pack of 200
30041461 2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker 55.00 mm Polypropylene Pack of 400
30041411 2 Stacked Red Diamonds Plus 2 Additional Red Diamons per Sticker 55.00 mm Paper Pack of 400
30041412 3 Red Diamonds per Sticker 105.00 mm Paper Pack of 200
30041476 3 Stacked Red Diamonds in Pyramid Formation per Sticker 105.00 mm Paper Pack of 200
30041452 3 Stacked Red Diamonds in Pyramid Formation per Sticker 55 mm Paper Pack of 400
30041414 2 Stacked Red Diamonds per Sticker 55 mm Polypropylene Pack of 400
30041453 3 Red Diamonds per Sticker in Rotated Pyramidal Shape 55 mm Polypropylene Pack of 400
30041464 4 Red Diamonds in Square Formation per Sticker 55.00 mm Polypropylene Pack of 400
30041477 3 Red Diamonds per Sticker in Rotated Pyramidal Shape 75.00 mm Polypropylene Pack of 300
30041424 4 Red Diamonds in Square Formation per Sticker 105.00 mm Polypropylene Pack of 200
30041465 3 Red Diamonds per Sticker in Rotated Pyramidal Shape 105.00 mm Polypropylene Pack of 200
30041446 1 Red Diamond per Sticker 55.00 mm Paper Pack of 400
30041480 4 Red Diamonds in Square Formation per Sticker 75.00 mm Polypropylene Pack of 300
30041455 2 Horizontal Red Diamonds per Sticker 105.00 mm Polypropylene Pack of 200
30041482 1 Red Diamond per Sticker 105.00 mm Polypropylene Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30041449 Leverantör Brady™ Leverantörsnummer 622307
  • ONLINE EXCLUSIVE PRICE
    138.00 €
    131.86 € / Pack of 400
    Save 6.14 €
    4% Off
Estimated Shipment: 30-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.

For you to print the required warning, compliant to the latest regulations.

Specifikationer

Certifications/Compliance UL Recognized
Description Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Gloss
Format Die-cut
Material Polypropylene
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Display Text 1 Red Diamond per Sticker
For Use With (Application) Hazard communication identification
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model R6000HF
No. per Pack 400 Labels/Roll
Quantity 400 Stickers
Shape Rectangle
Size 55.00 mm (W) x 85.00 mm (H)
Temperature Range (English) -4°F to 212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Dimensions (L x W) 55.00 mm (W) x 85.00 mm (H)
Length (English) 3.35 in.
Length (Metric) 85.00 mm
Width (English) 2.17 in.
Width (Metric) 55.00 mm
Width (English) Printable Area 55.00 mm
Width (Metric) Tape 55.00 mm
Height (English) 3.35 in.
Height (Metric) 85.00 mm
Alternate Part Number THTCLP-S-L1-7594-0.4-SC
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.