missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Small cupboard, red, 420 x 410 x 430mm
Description
All steel construction drawers for below workbench. Lockable cupboard and drawers, giving secured storage.
- Red
- Maximum load per drawer: 25kg
- Dimensions (H x W x D): 420mm x 410mm x 430mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction