Learn More
CELLPATH SMALL SLIDE BOX + SLEEVE (FLATPACK)
Product Code. 30226312
Change view
Click to view available options
Color:
Brown
Unit Size:
Pack of 25
Product Code. Color Unit Size
30226312 Brown Pack of 25
1 options
Product Code. 30226312 Supplier CELLPATH Supplier No. WES060008F
  • ONLINE EXCLUSIVE PRICE
    441.61 €
    184.84 € / Pack of 25
    Save 256.77 €
    58% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The robust Small Slide Box with Sleeve is suitable for archiving slides.
  • Key Benefits:
  • Made from recycled material
  • Bespoke boxes available at customer request
  • Cost efficient archive and storage
  • Dimension: 240 x 215 x 85 - holds 790 slides
Specifications
Type Slide Storage Box
Slide Capacity 790
Color Brown