missing translation for 'onlineSavingsMsg'
Learn More

Small Volume Sample Cups, Clarified Polypropylene, Conical Bottom, 13 x 25 mm (1.5 - 2.0 mL), 1000/pack

Product Code. 30317072
Change view
Click to view available options
Diameter (Metric) Outside:
13 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside
30317072 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317072 Supplier VHG Labs Supplier No. VHGFAASC3MP
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outside 13 mm
Quantity 1000/Pk.
Capacity (Metric) 1.5 - 2.0 mL
Material Clarified Polypropylene
Height (Metric) 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.