Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A
Shop All Thermo Scientific ProductsDescription
Use this kit to for purification of immunoglobulins prior to protein A digestion.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.
The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
Specifications
| Description | Magnetic, with SOLAμ Collection Plate, Soluble |
| Capture | Protein A |
| Cleavage | C-terminal side of lysine and arginine amino acids |
| Enzyme | Trypsin |
| Format | Bulk Magnetic |
| For Use With (Application) | Immunocapture and Protein Digest Sample Preparation |
| Product Line | SMART Digest |
| Type | Bulk Fractionation Kit |
| Includes | Bulk Magnetic SMART Digest Soluble Resin, Collection Plate, SOLAμ SPE Plate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.