missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A

Product Code. 15890172 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Magnetic, Soluble
Magnetic, Standalone
Magnetic, with SOLAμ Collection Plate, Soluble
Non-Magnetic, Soluble
Non-Magnetic, with SOLAμ Collection Plate, Soluble
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15890172 Non-Magnetic, Soluble Each
15800182 Non-Magnetic, with SOLAμ Collection Plate, Soluble Each
15810182 Magnetic, with SOLAμ Collection Plate, Soluble Each
15820182 Magnetic, Soluble Each
15870182 Magnetic, Standalone Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15890172 Supplier Thermo Scientific™ Supplier No. 60114101
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this kit to for purification of immunoglobulins prior to protein A digestion.

Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.

The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.

TRUSTED_SUSTAINABILITY

Specifications

Description Non-Magnetic, Soluble
Capture Protein A
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Non-magnetic
For Use With (Application) Immunocapture and Protein Digest Sample Preparation
Product Line SMART Digest
Type Bulk Fractionation Kit
Includes Bulk Non-magnetic SMART Digest Soluble Resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.