Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G
Shop All Thermo Scientific ProductsDescription
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
Specifications
Specifications
| Description | Magnetic, with SOLAμ Collection Plate, Soluble |
| Capture | Protein G |
| Cleavage | C-terminal side of lysine and arginine amino acids |
| Enzyme | Trypsin |
| Format | Bulk Magnetic |
| For Use With (Application) | Immunocapture and Protein Digest Sample Preparation |
| Product Line | SMART Digest |
| Type | SMART Digest Bulk Fractionation Kit |
| Includes | Bulk Magnetic SMART Digest Soluble Resin, Collection Plate, SOLAμ SPE Plate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.