missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Protein A Kits

Product Code. 15618693
Change view
Click to view available options
Description:
Magnetic
Magnetic with SOLAμ SPE and collection plate
Non-magnetic
Non-magnetic with SOLAμ SPE and collection plate
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15618693 Magnetic Each
15688683 Non-magnetic Each
15698683 Non-magnetic with SOLAμ SPE and collection plate Each
15608693 Magnetic with SOLAμ SPE and collection plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15618693 Supplier Thermo Scientific™ Supplier No. 60111104
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well for your biomarker studies.

Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well using Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Protein A Kits. These kits were designed to remove the challenges often associated with the process of biomarker quantitation by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight. The SMART Digest IA Protein A Kits target purification of immunoglobulins due to the strong affinity for binding to the Fc portion of the molecule.

Solving the challenges of low-level quantitation in complex matrices

The need to identify low-level biomarkers is forcing the analytical community to deal with extremely high levels of analyte and sample complexity, which makes quantitation challenging.

As these proteins are often present at low levels in complex biological matrices, an immunoaffinity capture step is typically employed to purify the sample and increase sensitivity. This step is often followed by protein digestion. Current processes are often time consuming, multistep, and prone to irreproducibility.

Immunocapture and Protein Digestion in a Single Well

These IA kits deliver a process that is:

  • Fast
  • Simple
  • Highly reproducible
  • Sensitive
Unique Design Reduces Workflow to Hours

In SMART Digest IA Kits, the immunocapture reagents and the temperature-activated, thermally stable trypsin are co-immobilized onto a resin. Following enrichment, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions, resulting in a workflow which takes hours rather than overnight.

TRUSTED_SUSTAINABILITY

Specifications

Description Magnetic
Capture Protein A
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format SMART Digest Resin, Magnetic
For Use With (Application) Peptide Quantitation
Product Line SMART Digest
Type Immunocapture + Protein Digestion combined on a single bead
Includes Magnetic SMART Digest Resin
How compatible are the SMART Digest ImmunoAffinity (IA) Kits with detergents (e.g. CHAPS, OGS, TWEEN and RIPA)?

We have observed the following effects with detergents:
• CHAPS – ~ 30% reduction in digest efficiency
• OGS – No reduction in digestion efficiency
• TWEEN – No reduction in digestion efficiency
• RIPA – The addition of RIPA, for ribonuclease A digestion, results in a concentration-dependent effect, where initial enzyme inhibition is overcome by improved substrate solubilization at higher concentrations only. 20% reduction in digestion efficiency.

We recommend compensating for reduction in digestion efficiency by extending the digestion time accordingly.

How much urea can I use with the SMART Digest ImmunoAffinity (IA) Kits?

The SMART Digest kits are not affected by concentrations of up to 0.5M of urea. If the concentration is higher than this, we recommend that the sample is diluted to <0.5 M of urea, prior to beginning digestion using the SMART Digest kit.

What is the composition of the SMART Digest IA buffer that comes in the SMART Digest ImmunoAffinity (IA) Kit?

The composition of the SMART Digest IA buffer is as follows:
Chemical Name (CAS No., EINECS No.): Kit Component Qty, Weight %
• Water (7732-18-5, 231-791-2): 2, 50-95%
• Glycerol (56-81-5, 200-289-5): 2, < 20%
• Tris Base (77-86-1, 201-064-4): 2, < 10%
• Tris-HCI (1185-53-1, 214-684-5): 2, < 10%
• Calcium Chloride (10043-52-4, 233-140-8): 2, < 10%
• Sodium Azide (26628-22-8, 247-852-1): 2, < 0.1%

Why don't I need to quantitate the total amount of protein and titer the enzyme when using the SMART Digest ImmunoAffinity (IA) Kits?

The SMART Digest IA Kits contain an excess of enzyme capable of digesting between 200 pg and 3.5 mg of protein. As most samples will fall within this range, it is not necessary to routinely quantitate protein concentration prior to digestion.

Why don't I need to use urea to unfold the protein when using SMART Digest ImmunoAffinity (IA) Kits?

The reason urea is needed as a first step in an in-solution protocol is to disrupt the sample and partially unfold the proteins. The proteins need to be partially unfolded so that the trypsin enzyme can have better access to the internal amino acid chain, not just the surface of the protein of interest. The reason the SMART Digest IA kits do not need urea is that it uses heat to unfold the protein.

Can I use the SMART Digest ImmunoAffinity (IA) Kits with Lys-C or other enzymes?

Yes, perform additional enzymatic digest reactions after digestion with the SMART Digest ImmunoAffinity (IA) Kits.

Are the SMART Digest ImmunoAffinity (IA) Kits compatible with gels?

No, the kits are not compatible with gels as the beads are unable to penetrate gels and start digestion.

Are the SMART Digest ImmunoAffinity (IA) Kits compatible with isobaric tagging (e.g. SILAC, ITRAQ, etc.)?

The current digest buffer contains amines so it is not compatible with isobaric tagging. However, amine-free buffers are available. For more information contact Technical Support at www.thermofisher.com/chromexpert

Why do I need to perform reduction and alkylation post digestion when using SMART Digest ImmunoAffinity (IA) Kits?

The reducing agent lowers digestion efficiency and adds extra steps unless you are specifically looking for disulphides.

What is the resin made of that the trypsin is coupled to in the SMART Digest ImmunoAffinity (IA) Kits?

The resin is a 20 µm PS-DVB core made hydrophilic with a two-tailed coating.

Will my sample be completely digested with SMART Digest ImmunoAffinity (IA) Kits?

Yes, our extensive studies have shown that complete sample digestion is achieved in as little as 5 minutes for simple, mono-protein samples, and 3.5 hours for complex matrices, such as plasma.

How can I mitigate trypsin self-digestion (autolysis) when using SMART Digest ImmunoAffinity (IA) Kits?

The immobilization of trypsin in SMART Digest ImmunoAffinity (IA) Kits prevents it from attacking and digesting itself, contrary to what happens in an in-solution digestion.

Is the trypsin mutated to be heat stable in SMART Digest ImmunoAffinity (IA) Kits? Would this affect my sample?

During the immobilization process, the trypsin is chemically modified in such a way that it is chemically stabilized while maintaining its specificity. The selectivity of the cleavage site is not affected.

Can I use surfactants with SMART Digest ImmunoAffinity (IA) Kits?

Many surfactants negatively impact not only digestion, but LC-MS performance as well. Of the surfactants we have screened, octylglucoside is the only MS compatible surfactant that does not negatively impact trypsin activity. It is not charged, so it does not impact MS ionization. It also exists as one molecular weight, so it does not result in multiple background peaks.

If I perform the digestion using one of the SMART Digest ImmunoAffinity (IA) Kits at high temperatures, will it result in an increase in post-translational modifications (PTMs)?

In comparison to in-solution digests, a comparable number of PTMs have been observed when screening for deamidation, amidation, methylation and oxidation. No modifications to existing PTMs, such as phosphorylated sites, have been observed.

Will disulfide bonds scramble during digestion with the SMART Digest ImmunoAffinity (IA) Kits?

If there are free cysteines, it is possible for disulfides to scramble before, during or after digestion. We would therefore recommend performing alkylation prior to digestion.

Do I have to reduce and alkylate my protein when using the SMART Digest ImmunoAffinity (IA) Kits?

The SMART Digest IA kits were engineered to be thermally stable. When operated at high temperatures (e.g. 70°C), denaturation and digestion occurs simultaneously. Therefore, for many quantitative workflows, there is no need to perform the additional steps of denaturation, reduction and alkylation. However, during this process many disulfide bonds will remain intact. As a result for characterization workflows where maximum sequence coverage is required we recommend that you perform reduction and alkylation after digestion. Denaturants and reducing reagents can negatively impact digestion using the SMART Digest IA kits.

What is the pH of the SMART Digest IA buffer that comes in the SMART Digest ImmunoAffinity (IA) Kits?

The pH of the SMART Digest IA buffer is approximately 7.2.

Are there salts in the SMART Digest IA buffer that comes in the SMART Digest ImmunoAffinity (IA) Kits?

The SMART Digest buffer in the SMART Digest ImmunoAffinity (IA) Kits contains about 0.5M salts. These salts enable quick digestion at high temperatures. We recommend using valve switching or Thermo Scientific SOLAµ SPE cleanup for desalting.

Do I have to use the SMART Digest IA buffer that comes in the SMART Digest ImmunoAffinity (IA) Kits?

The SMART Digest IA buffer was optimized for maximum trypsin activity at elevated temperatures. Other buffers can be used, but their use may negatively impact trypsin activity. If your application requires the use of an alternative buffer, digestion time and temperature should be optimized accordingly.

How long should my samples take to digest if I am using one of the SMART Digest ImmunoAffinity (IA) Kits?

All proteins vary with regards to digestion. Temperature and incubation times can be adjusted for optimal results. We recommend the following process for screening digestion time of your protein:
1. Create a method in your heater/shaker and set the temperature to 70°C and RPM to 1400.
2. Allow the temperature to reach equilibrium for at least 5 minutes.
3. Prepare eight identical samples using a relatively high known concentration of native analyte in the matrix of operation. Dilute them to 50 µL with ultrapure water, if necessary. Add them to individual SMART Digest wells.
4. Add 150 µL of SMART Digest IA buffer to each well and cap.
5. Place all wells firmly into the pre-heated heater/shaker.
6. Periodically remove a sample from the strip (every 5 to 15 minutes).
7. Centrifuge, filter or perform an SPE process with a SOLAµ HRP plate (Cat. No. 60209-001) and then analyze the samples to determine the extent of digestion.

Recommended digestion time for 200 µL protein solution (100µg/mL) at 70°C:
(Protein - digestion time)
• Insulin - 4 minutes
• BSA - < 5 minutes
• Carbonic anhydrase - < 5 minutes
• Lysozyme - < 5 minutes
• Apo-B - 30 minutes
• IgG - 45 minutes
• IgG in 50 µL plasma - 75 minutes (17.5 µg/mL)
• Ribonuclease A - 150 minutes
• Thyroglobulin - 240 minutes
• C-reactive protein - 240 minutes

I am using one of the SMART Digest ImmunoAffinity (IA) Kits. Can I vary the amount of trypsin I use depending on the protein load?

One of the benefits of using immobilized trypsin is that there is reduced autolytic activity. As a result, there is no need to vary the amount of trypsin used for any given sample.

How much trypsin is there in each well for the SMART Digest ImmunoAffinity (IA) Kits?

Every 30 µL of beads contains 14 µg of immobilized trypsin.

What amount of materials can I digest with the SMART Digest ImmunoAffinity (IA) Kits?

We recommend a digesting material ranging from 200 pg to 3.5 mg.

What volume of materials can I digest post-enrichment with the SMART Digest ImmunoAffinity (IA) Kits?

Following the immunoaffinity enrichment and wash steps, the sample volume is generally reduced to 50 µL or less. For every 1 µL of sample remaining, 3 µL of digest buffer should be added. The minimum recommended digestion volume is 50 µL (12.5 µL sample post wash, 37.5 µL digestion buffer) and the maximum recommended volume is 400 µL. Generally, 200 µL is used (50 µL sample post wash and 150µL digestion buffer). Refer to the SMART Digest ImmunoAffinity (IA) Kit User Manual for additional information).

What kind of samples have you used with the SMART Digest ImmunoAffinity (IA) Kits?

We have successful applications in mouse, monkey, beagle and human plasmas. We also have successful applications in cell lysates, urine and cerebral spinal fluid.

Can I use my standard PCR instrument for the SMART Digest ImmunoAffinity (IA) Kits?

We do not recommend using your standard PCR instrument because the shaking is necessary to avoid any diffusion limitations.

What heater shaker device should I use with the SMART Digest ImmunoAffinity (IA) Kits?

Uniform heating is key to sample reproducibility; shaking is a necessity to avoid any diffusion limitations. A heater shaker device with the following features is required:
• Heating block capable of uniformly heating samples to 70 °C
• Heated lid
• Shaking

What volume of materials can I process per well with the SMART Digest ImmunoAffinity (IA) Kits?

Volumes ranging from 5 to 1000 µL of sample per well are readily processed.

Can I use the Protein A and/or G versions of the SMART Digest ImmunoAffinity (IA) Kits to perform enrichment of all IgGs present in the sample?

Samples can be added directly to the SMART Digest IA kit, Protein A and G beads for bulk fractionation of immunoglobulins. For this workflow, no cross-linking is required. Be sure to take into account the capacity of the beads when developing a bulk fractionation workflow.

What is the best method for cross-linking my antibody for the SMART Digest ImmunoAffinity (IA) Protein A and G Kits?

The literature shows that many crosslinking methods are effective. We recommend using a glutaraldehyde based cross-linking method. For additional details refer to the cross-linking procedure outlined in the SMART Digest ImmunoAffinity (IA) Kit User Manual.

Will I see a change in loading capacity for the SMART Digest ImmunoAffinity (IA) Kits with biotin or biotin-like compounds in plasma?

We did not observe any reduction in loading capacity in tests where the resin was prewashed with plasma, compared to those were it was loaded with antibody first.

Will my protein digest during the affinity step in the SMART Digest ImmunoAffinity (IA) Kits?

The digestion is temperature and buffer-dependent. Because the affinity step is performed at lower temperatures (room temperature), in an optimal binding/suboptimal digestion buffer, no digestion is normally observed during the affinity step. Once the buffer is changed to the digestion buffer, which is designed to enhance digestion, and the sample is heated, digestion will proceed rapidly.

What is the loading capacity of the beads in the SMART Digest ImmunoAffinity (IA) Kits?

We recommend using a 30 µL aliquot of beads per sample. The loading capacity for 30 µL of beads is at least 5 µg of antibody or more.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.