Learn More
Thermo Scientific™ Smart NIR Integrating Sphere Accessories
Product Code. 18923491 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Reference Sample
Sample Spinner
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
18923491 Sample Spinner Each
18933481 Reference Sample Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18923491 Supplier Thermo Scientific™ Supplier No. 840095700
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessories for the Thermo Scientific™ Smart NIR Integrating Sphere accessory.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Smart NIR Integrating Sphere
Type Sample Spinner
For Use With (Application) Inhomogenous samples
Includes 5cm open powder sampling cup
Optics Quartz window