Learn More
Ibidi Smart Slide
Product Code. 15955091
Encompass_Preferred
Change view
Click to view available options
Smart Slide
Product Code.
15955091 Smart Slide
1 options
Product Code. 15955091 Supplier Ibidi Supplier No. 80816
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Smart Slide is an 8 channel cell counting chamber compatible with the Roche Cedex XS Analyzer.
  • An 8 channel cell counting chamber compatible with the Roche Cedex XS Analyzer
  • 15 x 8 measurements
  • sterilized
Specifications
For Use With (Equipment) Roche Cedex XS Analyzer
Sterility Sterile
Type Microscope Slide
Well Count 8
Quantity 15