Learn More
LevGo™ SmartSlicer™ Safety Razor Blades
Product Code. 15326558
Change view
Click to view available options
Size:
Small
Large
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15326558 Large Pack of 100
15316558 Small Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15326558 Supplier LevGo™ Supplier No. 30203
  • ONLINE EXCLUSIVE PRICE
    33.62 €
    27.15 € / Pack of 100
    Save 6.47 €
    19% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Say no to dangerous sharps. SmartSlicer™ Safety Razor Blades won't scratch surfaces when cutting agarose gels.
  • Cleanly slice agarose gels* right on the light box without scratching the surface.
  • Ideal for cutting soft tissue and plant matter.
  • Adjust the size required by simply cutting with scissors.
  • Available in two sizes and made in the USA from food grade PET.
  • Supplied in packs of 100.
Specifications
Blade Material Food Grade PET
Product Type Blade
For Use With (Application) Slicing Agarose Gels, Cutting Soft Tissue and Plant Matter
Size Large
Disposable Single-use
Sterility Non-sterile
Thickness (Metric) 0.22 mm
Certifications/Compliance Food Grade
Autoclavable Not Autoclavable
Quantity 100 Pk.
Graduated No