missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SMBA 1
GREENER_CHOICE

Product Code. 18736238
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18736238

Brand: Tocris Bioscience™ 5314/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and selective activator of Bax

SMBA 1 is a high affinity and selective activator of Bax (Ki = 43.3 nM). Binds to the S184 binding pocket to block Bax phosphorylation. Does not bind to other Bcl-2 family members, including Bcl-2, Bak and Bid. Facilitates Bax insertion into the mitochondrial membrane and promotes Bax-dependent cytochrome c release. Induces apoptosis in lung cancer cell lines expressing high levels of Bax selectively over normal cells expressing low levels of Bax. Represses tumor growth of A549 lung cancer xenografts in mice.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type SMBA 1
Purity >98%
Molecular Weight (g/mol) 315.32
Inhibitors SMBA 1

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.