missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SMER 3

Product Code. 18035012
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18035012 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18035012 Supplier R&D Systems™ Supplier No. 4375/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of E3 ubiquitin ligase

Selective inhibitor of a yeast SCF family E3 ubiquitin ligase (SCFMet30) in vitro and in vivo. Induces the expression of MET genes; blocks cell proliferation. Enhancer of rapamycin (Cat. No. 1292).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67200-34-4
Molecular Formula C11H4N4O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 224.18
Purity >98%
Molecular Weight (g/mol) 224.18
Chemical Name or Material SMER 3
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.