Learn More
Motic™ SMZ-160 Binocular Head
Product Code. 17867285
Change view
Click to view available options
Head Style:
Binocular
Unit Size:
Each
Product Code. Head Style unitSize
17867285 Binocular Each
1 options
Product Code. 17867285 Supplier Motic™ Supplier No. 1101010100911
Only null left
Add to basket
Add to basket
This Binocular Head is compatible with the SMZ-160 microscope.
  • SMZ-160-Binocular Head.
  • Includes 2 10x/20 mm eyepieces.
Specifications
Includes 2 eyepieces
Magnification Power 10X
Objective Types None
Field of View 20 mm
For Use With (Equipment) Head only, need also a stand and light
Warranty 5 years
Product Type Binocular head
Model SMZ-160
Head Style Binocular
Lighting Type None
For Use With (Application) Observation of sample in 3D
Eyepiece 10X/20 mm
Quantity 1