missing translation for 'onlineSavingsMsg'
Learn More

Motic™ SMZ 160 Stereo Microscopes

Product Code. 17860015
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Base Style:
LED Stand
Plain Stand
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Head Style Base Style Unit Size
17860015 Trinocular LED Stand Each
17847285 Binocular Plain Stand Each
17826011 Binocular LED Stand Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17860015 Supplier Motic™ Supplier No. 1100201300181
  • ONLINE EXCLUSIVE PRICE
    993.90 €
    839.09 € / Pack of 1
    Save 154.81 €
    15% Off
View all promotions
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This stereo zoom microscope comes with a 1:6.4 zoom range and an all metal body with classic illumination.

The SMZ-160 is ideal for educational purposes in high schools and universities, but is also great for industrial quality control.

  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6:1 zoom ratio
  • Diopter Adjustment: ±5

Specifications

Base Style LED Stand
Magnification Power 10X
Objective Types Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X)
Head Style Trinocular
Lighting Type R2 LED
Eyepiece WF 10X/20 mm
Working Distance 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.