Learn More
Motic™ SMZ-160-Trinocular head
Product Code. 17857285
Change view
Click to view available options
Head Style:
Trinocular
Unit Size:
Each
Product Code. Head Style Unit Size
17857285 Trinocular Each
1 options
Product Code. 17857285 Supplier Motic™ Supplier No. 1101010100921
  • ONLINE EXCLUSIVE PRICE
    609.00 €
    532.81 € / Pack of 1
    Save 76.19 €
    12% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This Trinocular Head is compatible with the SMZ-160 microscope.
  • SMZ-160-Trinocular Head.
  • Includes 2 10x/20 mm eyepieces.
Specifications
Includes 2 eyepieces of 10X/20 mm
Magnification Power 10X
Objective Types None
Field of View 20 mm
For Use With (Equipment) Head only, need also a stand and light
Warranty 5 years
Model SMZ-160 (Trinocular Head only)
Head Style Trinocular
Lighting Type None
For Use With (Application) Observation of Sample in 3D
Eyepiece 10x/20 mm