missing translation for 'onlineSavingsMsg'
Learn More

sn-Glycero-3-phosphocholine, MedChemExpress

Product Code. 16429867
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429867 5 g 5g
16401699 1 g 1g
16417398 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16429867 Supplier Medchem Express Supplier No. HY17552
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.

sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.

Specifications

Chemical Name or Material sn-Glycero-3-phosphocholine
CAS 28319-77-9
Color White
Purity Grade Notes Research
Molecular Formula C8H20NO6P
Quantity 5 g
Synonym Choline Alfoscerate Alpha-GPC L-α-GPC
Solubility Information H2O : 500 mg/mL (1943.86 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (388.77 mM; Need ultrasonic)
SMILES O=P(OCC[N+](C)(C)C)(OC[C@H](O)CO)[O-]
Molecular Weight (g/mol) 257.22
Formula Weight 257.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.