missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SNAP

Product Code. 30001104
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001104 50 mg 50mg
18045862 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001104 Supplier R&D Systems™ Supplier No. 0598/50
  • ONLINE EXCLUSIVE PRICE
    457.33 €
    409.50 € / 50mg
    Save 47.83 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. A stable analog of endogenous S-nitroso compounds

A stable analog of endogenous S-nitroso compounds. A source of NO in vivo which unlike organic O-nitrates does not induce tolerance. Decomposes slowly in solution with a t½ of 37h.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79032-48-7
Molecular Formula C7H12N2O4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 220.24
Purity >98%
Molecular Weight (g/mol) 220.24
Chemical Name or Material SNAP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.