missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 1.5 mL Flex-Tube™ - Microtube

Product Code. 13094697 Shop All Eppendorf Products
Click to view available options
Color:
Clear
Purity or Quality Grade:
Eppendorf Quality
Unit Size:
Pack of 500
This item is not returnable. View return policy

Product Code. 13094697

Brand: Eppendorf™ 022364111

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Flex-Tube™ (1.5 mL) are designed for seamless one-handed operation with an easy-to-open, securely closing tube design. Ideal for sample preparation, centrifugation, and storage, these tubes will effortlessly integrate into your workflows for efficient and reliable performance.

  • Easy-to-use ergonomic design featuring a user-friendly tube lid.
  • Durable polypropylene construction capable of withstanding chemicals, mechanical stress, and extreme temperatures.
  • g-Safe™ technology ensures stability up to 30,000 x g, preventing sample loss during centrifugation and enhancing safety with hazardous samples.
  • Free from slip agents, plasticizers, and biocides, eliminating substances that may compromise bio-assay results.
  • Clear graduation marks allow for visual volume verification during pipetting.
  • Convenient frosted lid and side surface facilitate easy labeling.
  • Functional across a wide temperature range (-86 °C to 100 °C) for versatile applications.
  • Autoclavable up to 121 °C for 20 minutes, ensuring effective sterilization.
  • Available in both Eppendorf Quality and PCR Clean purity grades with lot certification.

Spezifikation

Cap Style Snap Cap
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
For Use With (Application) For centrifuging or storing samples
Includes Flex lid
No. per Pack 500 Pk.
Packaging 1 Bag x 500 Tubes
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Capacity (English) 0.05 oz.
Closure Type Snap Cap
Length (English) 1.5 in.
Bottom Shape Conical
Max. RCF 30,000 x g
Graduations 0.5 mL
Diameter (Metric) 10 mm
Quantity 500 Tubes
Shape Cylindrical
Length (Metric) 38 mm
Product Type Microtube
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt