missing translation for 'onlineSavingsMsg'
Learn More

WHEATON™ Snap/Clip Top Closure for Vials

Product Code. 30286505 Shop All Wheaton Science Products Products
Change view
Click to view available options
Quantity:
140/Pk.
162/Pk.
190/Pk.
Unit Size:
Pack of 140
Pack of 162
Pack of 190
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30286505 162/Pk. Pack of 162
30286512 140/Pk. Pack of 140
30286509 190/Pk. Pack of 190
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30286505 Supplier WHEATON™ Supplier No. 105050162
  • ONLINE EXCLUSIVE PRICE
    11.94 €
    10.59 € / Pack of 162
    Save 1.35 €
    11% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Snap/Clip Top Vial Closures

  • WHEATON™ Snap/Clip Top Vials are manufactured from clear 5.1 expansion borosilicate glass and use the WHEATON™ 22 mm LDPE Snap/Clip Closure
  • Both the vials and caps are particularly suitable for the storage of powders and specimens due to the wide neck opening
  • The vials have a snap ring neck finish for the 'flip off' style 22 mm flexible low density polyethylene snap cap
  • Snap/Clip Top vials are available in sizes from 7 mL to 30 mL to allow flexibility in your sample storage
  • 22 mm LDPE snap caps are sold separately to the vials, and the one size fits all the vial sizes
  • The snap caps are sold in packs of 140, 162 and 190 to match vial quantities

Specifications

Color Natural
Diameter (Metric) Outer 22 mm
Material Polyethylene
Pre-slit No
Sterility Non-sterile
Quantity 162/Pk.
For Use With (Equipment) Vial
Autoclavable Not Autoclavable
Product Type Snap Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.