Learn More
Witeg™ Soda Glass Evaporating Dish
Description
- Watch glass shape
- Thin glass
- Fire-polished rim
- Soda glass
Specifications
Specifications
| Material | Soda Lime Glass |
| Shape | Round |
| Type | Evaporating Dish |
| Interior Finish | Polished |
| Diameter (Metric) | 70 mm |